Learn More
Vici Valco Instruments™ PEEK Union
Click to view available options
Type:
Union
Union with Fittings
Unit Size:
Each
Description
- Easy connection of transfer/solvent lines
- Low dead volume
- Completely inert and biocompatible
Specifications
Specifications
| Type | Union with Fittings |
| Diameter (Metric) Inner | 0.25 mm |
| Packaging Quantity | 1 Each |
| Material | PEEK |