missing translation for 'onlineSavingsMsg'
Learn More

Vici Valco Instruments™ PEEK Tubing

Product Code. 13113573
Change view
Click to view available options
Diameter (Metric) Inner:
0.13 mm
0.75 mm
Color:
Green
Red
Unit Size:
3m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Color Unit Size
13113573 0.75 mm Green 3m
13173563 0.13 mm Red 3m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13113573 Supplier Vici Valco Instruments™ Supplier No. 724JRT6003M3
  • ONLINE EXCLUSIVE PRICE
    £117.60
    £20.77 / 3m
    Save £96.83
    82% Off
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Color-coded PEEK tubing strong enough for continuous use at HPLC pressure without swelling or bursting. Vici Valco Instruments™ PEEK Tubing is not affected by halide salts, high-strength buffers, or other aggressive mobile phases that corrode stainless steel.

  • Color-coded based on size (interior diameter) and length
  • Strong enough to withstand continuous use at HPLC pressure without swelling or bursting
  • Not affected by halide salts, high strength buffers, or other aggressive mobile phases that corrode stainless steel
  • Polymer surface will not leach metal ions into the eluent
  • Polymer surface will not extract metal-sensitive components from the sample
  • Note: dichloromethane, THF, and DMSO may cause swelling in PEEK, and concentrated nitric and sulphuric acid will attack PEEK.

Specifications

Type Tubing
Color Green
For Use With (Application) Chromatography
Diameter (English) Outer 1/16 in.
Diameter (Metric) Inner 0.75 mm
Material PEEK
Max. Operating Temperature (Metric) 100°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.