Learn More
Trajan Scientific and Medical PEEK Tubing, 1/16 in. O.D., 100 ft
Designed specifically for modern LC, LCMS and automation useage. A flexible alternative to stainless steel tubing in high pressure applications, it ensures reproducibility in your chromatography flow rates and results through smooth internal surface along with consistent and tight toleranced IDs
Marca: Trajan Scientific and Medical 1302030005-100F
Dettagli aggiuntivi : Weight : 0.50000kg
Descrizione
- Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
- Bio-inert ad bio-compatible
- Color coded to industry standards for easy identification of inner diameter
- Leach-free colorants
- Smooth inner walls and precise IDs
- Works well with either polymer fittings as well as stainless steel fittings
- Can be formed to specific customer needs
Specifica
Green | |
0.030 in. | |
100 ft. | |
4000 psi (276 bar) |
2 to 14 | |
1/16 in. | |
PEEK | |
100°C |
For research use only