missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical PEEK Tubing, 1/16 in. O.D., 10 ft
Description
- Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
- Bio-inert ad bio-compatible
- Color coded to industry standards for easy identification of inner diameter
- Leach-free colorants
- Smooth inner walls and precise IDs
- Works well with either polymer fittings as well as stainless steel fittings
- Can be formed to specific customer needs
Specifications
Specifications
| Color | Yellow |
| pH Range | 2 to 14 |
| Diameter (English) Inner | 0.007 in. |
| Diameter (English) Outer | 1/16 in. |
| Overall Length (English) | 10 ft. |
| Material | PEEK |
| Maximum Pressure | 7000 psi (483 bar) |
| Max. Operating Temperature (Metric) | 100°C |
For research use only
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?