missing translation for 'onlineSavingsMsg'
Learn More

Vici Valco Instruments™ Peek Sample Loops

Product Code. 12700924
Click to view available options
Volume (Metric):
5 μL
50 μL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12700924

Brand: Vici Valco Instruments™ 724JRSLR2194

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Well suited for separations involving proteins, peptides, nucleic acids or other samples of biological origin as well as in IC systems.

Vici Valco Instruments™ Peek Sample Loops are made of Peek which is a mechanically strong, chemically inert polymer ideal for HPLC applications. Since our Peek sample loops contain no metals or other extractable compounds, no foreign materials will be introduced into the system. Softener-free polyethylene caps are used to avoid contamination of the tubing.

  • Fits all Rheodyne metal and metal-free valves
  • Wide range of volumes available
  • Biocompatible
  • PEEK sample loops are made from PEEK tubing and fittings.
  • Tolerances: ±0.025 mm

Specifications

Max. Pressure 400 bar
Volume (Metric) 5 μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.