Learn More
Vici Valco Instruments™ One-Piece Fingertight Column Coupler
Choose from a variety of coupler IDs, indicated by the color of the sleeve
Brand: Vici Valco Instruments™ JR-26504
Code : 15
Additional Details : Weight : 1.00000kg
Description
- Self-adjusting - zero dead volume for all fitting types
- Four bore sizes
- Biocompatible
- Adapts automatically to fit all pilot lengths
Specifications
Column Coupler | |
Orange | |
0.5 mm | |
PEEK |