Learn More
Cytiva PEEK Male Connector
Connect tubing of different sizes. Cytiva Life Sciences™ PEEK Male Connector is an important accessory for optimal chromatography.
Brand:  Cytiva 18-1120-90
Code : GE
Additional Details : Weight : 0.00376kg
 View more versions of this product
Product Code. 12421110
  • £112.00 / 1 set
Estimated Shipment: 05-06-2024
to see stock.
Add to basket
Description
Description
  • Chromatography accessory
  • Complete Packsize 2
  • Select from a variety of connections sizes and internal diamters 
  • For use with chromatography tubing
  • Connectors made of Polyetheretherketone (PEEK)
Specifications
Specifications
Male Connector
1/16 in. Male to 1/16 in. Male for tubing ID 0.13 mm
PEEK
0.13 mm