Learn More
Cytiva PEEK Male Connector
Product Code. 11515724 Shop All Cytiva Products
Change view
Click to view available options
Connections:
1/16 in. Female to M6 Male
1/16 in. Male
1/16 in. Male to 1/16 in. Male for tubing ID 0.13 mm
1/16 in. Male to 1/16 in. Male for tubing ID 0.25 mm
3 x 5/16 in. Female
3/16 in. Male
M6 Female to 5/16 in. Male
Unit Size:
1 set
Each
Pack of 10
Pack of 2
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connections unitSize
11515724 1/16 in. Female to M6 Male Pack of 2
11340062 1/16 in. Male Pack of 10
11729565 1/16 in. Male to 1/16 in. Male for tubing ID 0.25 mm Pack of 2
11370052 3/16 in. Male Each
12421110 1/16 in. Male to 1/16 in. Male for tubing ID 0.13 mm 1 set
11300072 3 x 5/16 in. Female Each
11360062 M6 Female to 5/16 in. Male Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11515724 Supplier Cytiva Supplier No. 18111257
In Stock
Add to basket
Add to basket
Connect tubing of different sizes. Cytiva Life Sciences™ PEEK Male Connector is an important accessory for optimal chromatography.
  • Chromatography accessory
  • Complete Packsize 2
  • Select from a variety of connections sizes and internal diamters
  • For use with chromatography tubing
  • Connectors made of Polyetheretherketone (PEEK)
TRUSTED_SUSTAINABILITY
Specifications
Type Male Connector
Connections 1/16 in. Female to M6 Male
Material PEEK