Learn More
Cytiva PEEK/ETFE Tubing Kit
Product Code. 12491120 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric) Outer:
1.58 mm
Unit Size:
1 set
Product Code. Diameter (Metric) Outer unitSize
12491120 1.58 mm 1 set
1 options
Product Code. 12491120 Supplier Cytiva Supplier No. 18112214
In Stock
Add to basket
Add to basket
Specifications
For Use With Classic AKTAexplorer
Type Tubing Kit
Includes PEEK 1 x Tubing G1, PEEK 1 x Tubing G2, PEEK 1 x Tubing G3, PEEK 1 x Tubing G4, PEEK 1 x Tubing G5, PEEK 1 x Tubing G6, PEEK 1 x Tubing G7, PEEK 1 x Tubing G8, PEEK 1 x Tubing G9, PEEK 1 x Tubing G10, EFTE 1 x Tubing G11, EFTE 1 x Tubing G12, EFTE 1 x Tubing G13, EFTE 1 x Tubing G14, EFTE 1 x Tubing G15, EFTE 1 x Tubing W3, 1/6 in. PEEK 40 x Finger tight Connector
Diameter (Metric) Outer 1.58 mm
Material ETFE/PEEK