missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ PEEK Analytical Guard Column Starter Kit

Product Code. 11543354
Change view
Click to view available options
Stationary Phase:
PRP-X110
PRP-X500
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Stationary Phase unitSize
11543354 PRP-X110 Each
11573354 PRP-X500 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11543354 Supplier Hamilton™ Supplier No. 79727
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

High sensitivity detection ion exchange chromatography of organic and inorganic anions, organic acids, organic and inorganic arsenic species, halides, nucleotides.

  • Separation Mode: Anion Exchange
  • Guard Cartridge Kit

Specifications

Particle Size 12 to 20 μm
Length (Metric) 8 mm
Max. Pressure 5000 psi
Diameter (Metric) 3 mm
Includes 1 holder, 2 cartridges
Stationary Phase PRP-X110
Material PEEK
Packing Material PSDVB/Trimethylammonium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.