missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Peek Analytical Guard Column Replacement Cartridges

Product Code. 11553394
Change view
Click to view available options
Packing Material:
PSDVB/Itaconate
PSDVB/Poly (dimethyl amido-propyl methacrylamide)
PSDVB/Trimethylammonium
Poly(methacryl amido-propyl trimethyl ammonium chloride)
Unit Size:
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packing Material unitSize
11553394 Poly(methacryl amido-propyl trimethyl ammonium chloride) Pack of 5
11533354 PSDVB/Itaconate Pack of 5
11513374 PSDVB/Poly (dimethyl amido-propyl methacrylamide) Pack of 5
12644885 PSDVB/Trimethylammonium Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11553394 Supplier Hamilton™ Supplier No. 79320
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Quantity 5 Pack
Particle Size 12 to 20 μm
Length (Metric) 8 mm
Max. Pressure 5000 psi
Type Column Replacement Cartridges
Diameter (Metric) Inner 3 mm
Stationary Phase PRP-X500
Material PEEK
Packing Material Poly(methacryl amido-propyl trimethyl ammonium chloride)
Column Format Replacement Guard Cartridge
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.