missing translation for 'onlineSavingsMsg'
Learn More

PEDFR/PNPLA2/ATGL Antibody [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30733575 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733575 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733575 Supplier Novus Biologicals Supplier No. AF5365JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

PEDFR/PNPLA2/ATGL Polyclonal antibody specifically detects PEDFR/PNPLA2/ATGL in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEDFR/PNPLA2/ATGL
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias Adipose triglyceride lipase, ATGL1110001C14Rik, Calcium-independent phospholipase A2, Desnutrin, EC 3.1.1.3, FP17548, IPLA2-zeta, patatin-like phospholipase domain containing 2, patatin-like phospholipase domain-containing protein 2, patatin-like phospholipase domain-containing protein 2-like, PEDF-R, Pigment epithelium-derived factor, Transport-secretion protein 2, transport-secretion protein 2.2, triglyceride hydrolase, TTS2.2, TTS-2.2, TTS2DKFZp667M109
Host Species Sheep
Immunogen E. coli-derived recombinant mouse PEDF R/PNPLA2, Val162-Gly253, Accession # Q8BJ56
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Diabetes Research, Lipid and Metabolism, Lipid Droplets, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57104
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.