missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEDF Mouse anti-Human, Porcine, Clone: 2C5C8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66554-1-IG-20UL
This item is not returnable.
View return policy
Description
The PEDF gene is a member of the serpin gene family. Serpins are a group of serine protease inhibitors, some of which have also been reported to exhibit neurotrophic activity.Specifications
| PEDF | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P36955 | |
| SERPINF1 | |
| PEDF Fusion Protein Ag26660 | |
| 20 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 2C5C8 | |
| Unconjugated | |
| SERPINF1 | |
| EPC 1, PEDF, PIG35, Serpin F1, SERPINF1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 5176, 780402 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction