missing translation for 'onlineSavingsMsg'
Learn More

PEDF Mouse anti-Human, Porcine, Clone: 2C5C8, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66554-1-IG-150UL

 View more versions of this product

Product Code. 16878535

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The PEDF gene is a member of the serpin gene family. Serpins are a group of serine protease inhibitors, some of which have also been reported to exhibit neurotrophic activity.
Specifications

Specifications

PEDF
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P36955
SERPINF1
PEDF Fusion Protein Ag26660
150 μL
Primary
Human, Pig
Antibody
IgG1
Western Blot
2C5C8
Unconjugated
SERPINF1
EPC 1, PEDF, PIG35, Serpin F1, SERPINF1
Mouse
Protein G
RUO
5176, 780402
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.