missing translation for 'onlineSavingsMsg'
Learn More

Pectin Citrus

Product Code. 15423689
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15423689 100 g 100g
15433689 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15423689 Supplier Thermo Scientific Alfa Aesar Supplier No. J61021.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9000-69-5

Pectin Citrus is promoted in dietary supplement form as an alternative cancer treatment. It can also been used to treat prostate cancer and melanoma. It can also be used as a gelling agent, filler, and stabilizer. It Supports celluar health, promotes immune function, supports cardiovascular health, supports healthy function of multiple organs, Safely chelates heavy metals and toxins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(RUO – Research Use Only.) Promoted in dietary supplement form as an alternative cancer treatment, Pectin Citrus can also been used to treat prostate cancer and melanoma. It can also be used as a gelling agent, filler, and stabilizer. It Supports celluar health, promotes immune function, supports cardiovascular health, supports healthy function of multiple organs, Safely chelates heavy metals and toxins.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9000-69-5
MDL Number MFCD00081838
Synonym Poly-D-galacturonic acid methyl ester

Specifications

Color Brown to Yellow
Quantity 100 g
Merck Index 14,7063
Solubility Information It is soluble in water.
Physical Form Powder
Chemical Name or Material Pectin Citrus

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.