missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pectin, pract.

Product Code. 10722412 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10722412 250 g Plastic bottle 250g
10279700 1 kg Plastic bottle 1kg
10452795 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10722412 Supplier Thermo Scientific Chemicals Supplier No. 416862500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9000-69-5

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9000-69-5
MDL Number MFCD00081838

Specifications

Color Orange to Orange-Red
pH 3.0 to 3.8 (1% soln.)
Physical Form Powder
Ash acid-insoluble ash: < 1%
Merck Index 15, 7169
Quantity 250 g
Percent Purity 67 to 71% (degree of esterification)
Grade Practical
Infrared Spectrum Authentic
Loss on Drying 12% max. (105°C, 2 hrs)
Packaging Plastic bottle
Particle Size 2% Max. (>60 Mesh)
Viscosity 30.5 to 36.5
Chemical Name or Material Pectin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.