missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PECR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522207
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PECR (Peroxisomal Trans-2-Enoyl-CoA Reductase) is a Protein Coding gene. Among its related pathways are Metabolism and Allograft rejection. Gene Ontology (GO) annotations related to this gene include signaling receptor binding and coenzyme binding. An important paralog of this gene is DECR1.
Specifications
| PECR | |
| Polyclonal | |
| Unconjugated | |
| Pecr | |
| 2,4-dienoyl-CoA reductase-related protein; 2400003B18Rik; DCRRP; DCR-RP; HPDHase; HSA250303; PECR; perosisomal 2-enoyl-CoA reductase; peroxisomal 2,4-dienoyl-CoA reductase; Peroxisomal trans-2-enoyl-CoA reductase; PRO1004; putative short chain alcohol dehydrogenase; PVIARL; pVI-ARL; PX-2,4-DCR1; RLF98; SDR29C1; Short chain dehydrogenase/reductase family 29C member 1; short chain dehydrogenase/reductase family 29C, member 1; TERP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55825 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9BY49 | |
| Pecr | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PECR. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction