missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PECI Monoclonal Antibody (K16256_1E9), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA556325

 View more versions of this product

Product Code. 30198225

  • £760.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: KDFENSMNQV KLLKKDPGNE VKLKLYALYK QATEGPCNMP KPGVFDLINK AKWDAWNALG SLPKEAARQN YVDLVSSLSP SLESSSQVEP GTDRKSTGFE TLVVTSEDGI TKIMFNRPKK KNAINTEMYH EIMRALKAAS KDDSIITVLT GNGDYYSSGN DLTNFTDIPP GGVEEKAKNN AVLLREFVGC FIDFPKPLIA VVNGPAVGIS VTLLGLFDAV YASDRATFHT PFSHLGQSPE GCSSYTFPKI MSPAKATEML IFGKKLTAGE ACAQGLVTEV FPDSTFQKEV WTRLKAFAKL PPNALRISKE VIRKREREKL HAVNAEECNV LQGRWLSDEC TNAVVNFLSR

PECI is an auxiliary enzyme that catalyzes an isomerization step required for the beta-oxidation of unsaturated fatty acids.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PECI
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
O75521
ECI2
Recombinant human ECI2 (immunogen range is 41-390).
200 μL
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
K16256_1E9
Unconjugated
ECI2
ACBD2; acyl-Coenzyme A binding domain containing 2; D3,D2-enoyl-CoA isomerase; DBI-related protein 1; delta(3),delta(2)-enoyl-CoA isomerase; Diazepam-binding inhibitor-related protein 1; dJ1013A10.3; Dodecenoyl-CoA isomerase; Drs1; DRS-1; Eci2; enoyl-CoA delta isomerase 2; enoyl-CoA delta isomerase 2, mitochondrial; enoyl-Coenzyme A delta isomerase 2; Hca88; Hepatocellular carcinoma-associated antigen 88; PECI; Peroxisomal 3,2-trans-enoyl-CoA isomerase; peroxisomal 3,2-trans-enoyl-CoA isomerase; enoyl-CoA delta isomerase 2, mitochondrial; peroxisomal D3,D2-enoyl-CoA isomerase; peroxisomal delta3, delta2-enoyl-Coenzyme A isomerase; renal carcinoma antigen NY-REN-1; testicular secretory protein Li 33
Mouse
Protein A
RUO
10455
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.