missing translation for 'onlineSavingsMsg'
Learn More

PECI Antibody (OTI2H10) [DyLight 350], Novus Biologicals™

Product Code. 30688642 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30688642 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688642 Supplier Novus Biologicals Supplier No. NBP344645UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PECI Monoclonal antibody specifically detects PECI in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PECI
Applications Western Blot
Classification Monoclonal
Clone OTI2H10
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias ACBD2, acyl-Coenzyme A binding domain containing 2, D2-enoyl-CoA isomerase, delta(2)-enoyl-CoA isomerase, Delta(3), Diazepam-binding inhibitor-related protein 1, dJ1013A10.3, Dodecenoyl-CoA isomerase, DRS-1, DRS1D3, EC 5.3.3.8, enoyl-CoA delta isomerase 2, HCA88DBI-related protein 1, Hepatocellular carcinoma-associated antigen 88, KIAA0536, PECI, Peroxisomal 3,2-trans-enoyl-CoA isomerase, peroxisomal D3, Renal carcinoma antigen NY-REN-1
Host Species Mouse
Immunogen Full length human recombinant protein of human PECI(NP_006108) produced in HEK293T cell.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 10455
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.