missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PECI Monoclonal antibody specifically detects PECI in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | PECI |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI1H4 |
| Conjugate | HRP |
| Formulation | PBS |
| Gene Alias | ACBD2, acyl-Coenzyme A binding domain containing 2, D2-enoyl-CoA isomerase, delta(2)-enoyl-CoA isomerase, Delta(3), Diazepam-binding inhibitor-related protein 1, dJ1013A10.3, Dodecenoyl-CoA isomerase, DRS-1, DRS1D3, EC 5.3.3.8, enoyl-CoA delta isomerase 2, HCA88DBI-related protein 1, Hepatocellular carcinoma-associated antigen 88, KIAA0536, PECI, Peroxisomal 3,2-trans-enoyl-CoA isomerase, peroxisomal D3, Renal carcinoma antigen NY-REN-1 |
| Host Species | Mouse |
| Immunogen | Full length human recombinant protein of human PECI(NP_006108) produced in HEK293T cell. |
| Purification Method | >95% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?