missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PECI Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35305-20ul
This item is not returnable.
View return policy
Description
PECI Polyclonal antibody specifically detects PECI in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| PECI | |
| Polyclonal | |
| Western Blot 1:1000 - 1:2000, ELISA | |
| ACBD2, acyl-Coenzyme A binding domain containing 2, D2-enoyl-CoA isomerase, delta(2)-enoyl-CoA isomerase, Delta(3), Diazepam-binding inhibitor-related protein 1, dJ1013A10.3, Dodecenoyl-CoA isomerase, DRS-1, DRS1D3, EC 5.3.3.8, enoyl-CoA delta isomerase 2, HCA88DBI-related protein 1, Hepatocellular carcinoma-associated antigen 88, KIAA0536, PECI, Peroxisomal 3,2-trans-enoyl-CoA isomerase, peroxisomal D3, Renal carcinoma antigen NY-REN-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-364 of human PECI (NP_006108.2).,, Sequence:, MNRTAMRASQKDFENSMNQVKLLKKDPGNEVKLKLYALYKQATEGPCNMPKPGVFDLINKAKWDAWNALGSLPKEAARQNYVDLVSSLSPSLESSSQVEPGTDRKSTGFETLVVTSEDGITKIMFNRPKKKNAINTEMYHEIMRALKAASKDDSIITVLTGNGDYYSSGNDLTNFTDIPPGGVEEKAKNNAVLLREFVGCFIDFPKPLIAVVNGPAVGISVTLLGLFDAVYASDRATFHTPFSHLGQSPEGCSSYTFPKIMSPAKATEMLIFGKKLTAGEACAQGLVTEVFPDSTFQKEVWTRLKAFAKLPPNALRISKEVIRKREREKLHAVNAEECNVLQGRWLSDECTNAVVNFLSRKSKL | |
| 20 μL | |
| Proteases & Other Enzymes | |
| 10455 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction