missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pecanex Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | Pecanex |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18207931
|
Novus Biologicals
NBP2-58037 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18665407
|
Novus Biologicals
NBP2-58037-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pecanex Polyclonal specifically detects Pecanex in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Pecanex | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 22990 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TGNNATTANNPHSNVTQGSIGNPGQGSGTGLHPPVTSYPPTLGTSHSSHSVQSGLVRQSPARASVASQSSYCYSSRHSSLRMSTTGF | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| FLJ23409, KIAA0805FLJ45663, KIAA0995pecanex-like 1, PCNXL1, Pecanex homolog, pecanex homolog (Drosophila), pecanex-like 1 (Drosophila), pecanex-like protein 1 | |
| PCNX1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title