missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PECAM1 Mouse anti-Human, PE, Clone: JC/70A, Abnova™
Mouse monoclonal antibody raised against native PECAM1.
Brand: Abnova MAB13014.500uL
This item is not returnable.
View return policy
Specifications
| PECAM1 | |
| Monoclonal | |
| PE | |
| Flow Cytometry (5 uL/million cells) Immunofluorescence (1:50-1:100 for 30 minutes at RT) | |
| CD31/PECAM-1 | |
| Mouse | |
| Protein A/G affinity chromatography | |
| RUO | |
| 5175 | |
| Store at 4°C. | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence | |
| JC/70A | |
| Mouse monoclonal antibody raised against native PECAM1. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| PECAM1 | |
| Membrane preparation of a spleen from a patient with hairy cell leukemia. | |
| 500 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction