Learn More
PEAR1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28086-1-AP-20UL
This item is not returnable.
View return policy
Description
The PEAR1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PEAR1. This antibody recognizes human antigen. The PEAR1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PEAR1 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 375033 | |
| IgG |
| Western Blot, ELISA | |
| 267 μg/mL | |
| Rabbit | |
| Primary | |
| Human |