missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEAR1 Polyclonal Antibody

PEAR1 Polyclonal Antibody
Brand: Invitrogen PA521057
This item is not returnable.
View return policy
Description
PEAR1 Polyclonal Antibody for Western Blot, IHC
Specifications
| PEAR1 | |
| Polyclonal | |
| Unconjugated | |
| PEAR1 | |
| 3110045G13Rik; hPEAR1; Jagged and Delta protein; Jedi; Jedi-1; MEGF12; mPEAR1; Multiple EGF-like domains protein 12; multiple EGF-like-domains 12; multiple epidermal growth factor-like domains protein 12; Pear1; Platelet endothelial aggregation receptor 1; protein Jedi; RGD1305653; RP11-107D16.3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 295293, 375033, 73182 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q5VY43, Q8VIK5 | |
| PEAR1 | |
| A 16 amino acid peptide near the carboxy terminus of human PEAR1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction