missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEAR1 Polyclonal Antibody

Description
Immunogen sequence: PSDPNTCSFW ESFTTTTKES HSRPFSLLPS EPCERPWEGP HTCPQPTVVY RTVYRQVVKT DHRQRLQCCH GFYES Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 79%.
Platelet endothelial aggregation receptor 1 (PEAR1) is a platelet receptor that signals upon the formation of platelet-platelet contacts independent of platelet activation and secondary to platelet aggregation. Upon platelet aggregation stimulated by physiological agonists, PEAR1 becomes tyrosine- and serine-phosphorylated; the tyrosine phosphorylation can be inhibited by eptifibatide, an aIIbbeta3 antagonist that also inhibits platelet aggregation. Recent studies have indicated that genetic variations in PEAR1, may be associated with enhanced agonist-induced platelet aggregation.
Specifications
Specifications
| Antigen | PEAR1 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | PEAR1 |
| Gene Accession No. | Q5VY43 |
| Gene Alias | 3110045G13Rik; hPEAR1; Jagged and Delta protein; Jedi; Jedi-1; MEGF12; mPEAR1; Multiple EGF-like domains protein 12; multiple EGF-like-domains 12; multiple epidermal growth factor-like domains protein 12; Pear1; Platelet endothelial aggregation receptor 1; protein Jedi; RGD1305653; RP11-107D16.3 |
| Gene Symbols | PEAR1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?