missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEAQX tetrasodium hydrate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
PEAQX (NVP-AAM077) tetrasodium hydrate is a potent, selective and orally active NMDA antagonist, with IC50 values of 270 nM and 29600 nM for hNMDAR 1A/2B and hNMDAR 1A/2B, respectively.
Specifications
Specifications
| Chemical Name or Material | PEAQX tetrasodium hydrate |
| Color | Earth Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C17H15BrN3Na4O6P |
| Quantity | 50 mg |
| Synonym | NVP-AAM077 tetrasodium hydrate |
| Solubility Information | H2O : 25.5 mg/mL (45.52 mM; Need ultrasonic and warming) |
| SMILES | BrC1=CC=C([C@H](C)NC(P(O[Na])(O[Na])=O)C2=CC=CC3=C2N=C(O[Na])C(O[Na])=N3)C=C1.[H]O[H] |
| Molecular Weight (g/mol) | 560.15 |
| Formula Weight | 560.15 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction