missing translation for 'onlineSavingsMsg'
Learn More

PEAQX tetrasodium hydrate, MedChemExpress

Product Code. 16427797
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16427797

Brand: Medchem Express HY12294A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PEAQX (NVP-AAM077) tetrasodium hydrate is a potent, selective and orally active NMDA antagonist, with IC50 values of 270 nM and 29600 nM for hNMDAR 1A/2B and hNMDAR 1A/2B, respectively.

PEAQX (NVP-AAM077) tetrasodium hydrate is a potent, selective and orally active NMDA antagonist, with IC50 values of 270 nM and 29600 nM for hNMDAR 1A/2B and hNMDAR 1A/2B, respectively.

Specifications

Chemical Name or Material PEAQX tetrasodium hydrate
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C17H15BrN3Na4O6P
Quantity 5 mg
Synonym NVP-AAM077 tetrasodium hydrate
Solubility Information H2O : 25.5 mg/mL (45.52 mM; Need ultrasonic and warming)
SMILES BrC1=CC=C([C@H](C)NC(P(O[Na])(O[Na])=O)C2=CC=CC3=C2N=C(O[Na])C(O[Na])=N3)C=C1.[H]O[H]
Molecular Weight (g/mol) 560.15
Formula Weight 560.15
Percent Purity 97.05%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.