missing translation for 'onlineSavingsMsg'
Learn More

Peanut Oil, MP Biomedicals

Product Code. 11442751 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 gal.
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442751 5 gal. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11442751 Supplier MP Biomedicals Supplier No. 0290468405
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 8005-03-6
Molecular Formula C22H14N6Na2O9S2
Quantity 5 gal.
Synonym amido black 10b, acid black 1, buffalo black nbr, naphthol blue black, aniline blue black, tech, nigrosine water soluble, 4-amino-5-hydroxy-3-4-nitrophenyl azo-6-phenylazo naphthalene-2,7-disulphonic acid, sodium salt, 2,7-naphthalenedisulfonic acid, 4-amino-5-hydroxy-3-4-nitrophenyl azo-6-phenylazo-, sodium salt, 3-4-nitrophenylazo-4-amino-5-oxo-6-2-phenylhydrazono-5,6-dihydro-2,7-naphthalenedisulfonic acid disodium salt, disodium 3e-5-amino-6-4-nitrophenyl diazenyl-4-oxo-3-phenylhydrazinylidene naphthalene-2,7-disulfonate
InChI Key HKBVRFLHNUEVRO-HMDKGIQMSA-L
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
Molecular Weight (g/mol) 616.49
PubChem CID 44134531
Chemical Name or Material Peanut Oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.