missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEAK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101940
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SGK269.
Tyrosine kinase that may play a role in cell spreading and migration on fibronectin. May directly or indirectly affect phosphorylation levels of cytoskeleton-associated proteins MAPK1/ERK and PXN.
Specifications
| PEAK1 | |
| Polyclonal | |
| Unconjugated | |
| PEAK1 | |
| 1110049L02Rik; 9530046P14; AA690169; BE132841; C230081A13Rik; Inactive tyrosine-protein kinase PEAK1; KIAA2002; mKIAA2002; NKF3 kinase family member; Peak1; pseudopodium enriched atypical kinase 1; pseudopodium-enriched atypical kinase 1; Sgk269; sugen kinase 269; Tyrosine-protein kinase SgK269 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 244895, 79834 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q69Z38, Q9H792 | |
| PEAK1 | |
| A synthesized peptide derived from human PEAK1(Accession Q9H792), corresponding to amino acid residues T539-N589. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction