missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PEAK1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA555302

Product Code. 15763221

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: DGQSICGELS IQEHCENKPV IIGWNRNRAA LSQKPLNNNN EDDEGISHVP KPYGNNDSAK KMSDNNNGLT EVLKEIAGLD TAPQIR Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 72%.

Tyrosine kinase that may play a role in cell spreading and migration on fibronectin. May directly or indirectly affect phosphorylation levels of cytoskeleton-associated proteins MAPK1/ERK and PXN.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PEAK1
Polyclonal
Unconjugated
PEAK1
1110049L02Rik; 9530046P14; AA690169; BE132841; C230081A13Rik; Inactive tyrosine-protein kinase PEAK1; KIAA2002; mKIAA2002; NKF3 kinase family member; Peak1; pseudopodium enriched atypical kinase 1; pseudopodium-enriched atypical kinase 1; Sgk269; sugen kinase 269; Tyrosine-protein kinase SgK269
Rabbit
Antigen affinity chromatography
RUO
79834
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9H792
PEAK1
Recombinant protein corresponding to Human PEAK1. Recombinant protein control fragment (Product #RP-95222).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.