missing translation for 'onlineSavingsMsg'
Learn More

Pea3 Antibody - BSA Free, Novus Biologicals™

Product Code. 18333810 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333810 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18333810 Supplier Novus Biologicals Supplier No. NBP281873

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Pea3 Polyclonal specifically detects Pea3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pea3
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA
Formulation PBS
Gene Alias Adenovirus E1A enhancer-binding protein, E1A-FE1A enhancer binding protein, E1AFETS translocation variant 4, ets variant 4, ets variant gene 4 (E1A enhancer binding protein, E1AF), ets variant gene 4 (E1A enhancer-binding protein, E1AF), EWS protein/E1A enhancer binding protein chimera, PEA3, PEAS3, Polyomavirus enhancer activator 3 homolog, polyomavirus enhancer activator-3, Protein PEA3
Gene Symbols ETV4
Host Species Rabbit
Immunogen Pea3 antibody was raised against a peptide corresponding to 19 amino acids near the amino terminus of human Pea3. Amino Acid Squence: HSPTTRIKKEPQSPRTDPA
Molecular Weight of Antigen 53 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2118
Test Specificity At least three isoforms of Pea3 are known to exist. This antibody will only detect the two longest isoforms.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.