missing translation for 'onlineSavingsMsg'
Learn More

PEA-15 Antibody, Novus Biologicals™

Mouse Monoclonal Antibody

£251.00 - £373.00

Specifications

Antigen PEA-15
Concentration 1 mg/mL
Dilution Western Blot 1:1000-1:2000, Immunocytochemistry/ Immunofluorescence 1:1000-2000
Applications Western Blot, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18620068
View Documents
Novus Biologicals
NBP3-05543-100ul
100 μg
£373.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18689756
View Documents
Novus Biologicals
NBP3-05543-50ul
50 μg
£251.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PEA-15 Monoclonal antibody specifically detects PEA-15 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

PEA-15
Western Blot 1:1000-1:2000, Immunocytochemistry/ Immunofluorescence 1:1000-2000
Monoclonal
Purified
RUO
Human, Mouse, Rat, Bovine
HMAT115kD, PEA-15, PED15 kDa phosphoprotein enriched in astrocytes, phosphoprotein enriched in astrocytes 15, Phosphoprotein enriched in diabetes
Full length human PEA-15 as expressed in and purified from E. coli.
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
1 mg/mL
Western Blot, Immunofluorescence
Unconjugated
Mouse
Apoptosis, Cell Biology, Cellular Markers
50% PBS, 50% glycerol
8682
IgG1
Immunogen affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.