missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDZRN4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93824-0.1ml
This item is not returnable.
View return policy
Description
PDZRN4 Polyclonal antibody specifically detects PDZRN4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PDZRN4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp434B0417, IMAGE5767589, Ligand of Numb protein X 4, LNX4FLJ33777, PDZ domain containing ring finger 4, PDZ domain-containing RING finger protein 4, SAMCAP3L, SEMACAP3-like protein, SEMCAP3L | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 220-450 of human PDZRN4 (NP_037509.3). RIVLLVARPEIQLDEGWLEDERNEFLEELNLEMLEEEHNEAMQPTANEVEQPKKQEEEEGTTDTATSSSNNHEKDSGVGRTDESLRNDESSEQENAAEDPNSTSLKSKRDLGQSQDTLGSVELQYNESLVSGEYIDSDCIGNPDEDCERFRQLLELKCKIRNHGEYDLYYSSSTIECNQGEQEGVEHELQLLNEELRNIELECQNIMQAHRLQKVTDQYGDIWTLHDGGFR | |
| 0.1 mL | |
| Zinc Finger | |
| 29951 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction