missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Research Category | Zinc Finger |
| Target | PDZRN3 |
| Synonym | E3 ubiquitin-protein ligase PDZRN3, EC 6.3.2.-, Ligand of Numb protein X 3, likely ortholog of mouse semaF cytoplasmic domain associated protein 3, LNX3KIAA1095SEMACAP3, PDZ domain containing ring finger 3, PDZ domain-containing RING finger protein 3, Protein SEMACAP3, Semaphorin cytoplasmic domain-associated protein 3, SEMCAP3 |
| Gene Symbol | PDZRN3 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?