missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDZK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10507-2-AP-150UL
This item is not returnable.
View return policy
Description
PDZK1 is a scaffold protein that connects plasma membrane proteins and regulatory components, regulating their surface expression in epithelial cells apical domains. It may be involved in the coordination of a diverse range of regulatory processes for ion transport and second messenger cascades. In complex with SLC9A3R1, it may cluster proteins that are functionally dependent in a mutual fashion and modulate the trafficking and the activity of the associated membrane proteins. It may also play a role in the cellular mechanisms associated with multidrug resistance through its interaction with ABCC2 and PDZK1IP1. May potentiate the CFTR chloride channel activity. PDZK1 functions to connect SCARB1 with the cellular machineries for intracellular cholesterol transport and/or metabolism and may be involved in the regulation of proximal tubular Na(+)-dependent inorganic phosphate cotransport therefore playing an important role in tubule function.Specifications
| PDZK1 | |
| Polyclonal | |
| Unconjugated | |
| PDZK1 | |
| CAP70, CLAMP, NaPi Cap1, NHERF 3, NHERF3, PDZ domain containing 1, PDZD1, PDZK1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5174, 59020, 65144 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5T2W1, Q9JIL4, Q9JJ40 | |
| Pdzk1 | |
| PDZK1 Fusion Protein Ag0787 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction