missing translation for 'onlineSavingsMsg'
Learn More

PDZK1 Antibody (SR2158), Novus Biologicals™

Rabbit Monoclonal Antibody

£257.00 - £341.00

Specifications

Antigen PDZK1
Clone SR2158
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18635634
View Documents
Novus Biologicals
NBP3-22197-100ul
100 μg
£341.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18643725
View Documents
Novus Biologicals
NBP3-22197-50ul
50 μg
£257.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PDZK1 Monoclonal antibody specifically detects PDZK1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications

Specifications

PDZK1
Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Monoclonal
Purified
RUO
Human
CAP70, CFTR-associated protein of 70 kDa, CLAMP, Na(+)/H(+) exchanger regulatory factor 3, Na/Pi cotransporter C-terminal-associated protein 1, naPi-Cap1, NHERF-3, NHERF3PDZD1Na(+)/H(+) exchange regulatory cofactor NHE-RF3, PDZ domain containing 1, PDZ domain-containing protein 1, PDZ-containing kidney protein 1, Sodium-hydrogen exchanger regulatory factor 3
A synthesized peptide derived from human PDZK1 (Uniprot #: Q5T2W1)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR2158
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Cellular Markers, Diabetes Research, Endothelial Cell Markers, Growth and Development, Lipid and Metabolism, Neuronal Cell Markers, Plasma Membrane Markers, Signal Transduction
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
5174
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.