missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDZD2/PDZK3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-33651
This item is not returnable.
View return policy
Description
PDZD2/PDZK3 Polyclonal specifically detects PDZD2/PDZK3 in Human samples. It is validated for Western Blot.
Specifications
| PDZD2/PDZK3 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| PDZD2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human PDZK3. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 23037 | |
| Human | |
| IgG |
| Western Blot | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000 | |
| Activated in prostate cancer protein, AIPCPAPIN, KIAA0300PDZ domain-containing protein 3, PDZ domain containing 2, PDZ domain containing 3, PDZ domain-containing protein 2, PDZK3, PIN1 | |
| Rabbit | |
| 302 kDa | |
| 100 μL | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction