missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDXP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551404
This item is not returnable.
View return policy
Description
Immunogen sequence: FAKLREACAH LRDPECLLVA TDRDPWHPLS DGSRTPGTGS LAAAVETASG RQALVVGKPS PYMFECITEN FSIDPARTLM VGDRLETDIL FGHRCGMTTV LTLTGVSRLE EAQAYLAAGQ HDLVPHYYVE SIADLTE Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.
Pyridoxal 5-prime-phosphate (PLP) is the active form of vitamin B6 that acts as a coenzyme in maintaining biochemical homeostasis. The preferred degradation route from PLP to 4-pyridoxic acid involves the dephosphorylation of PLP by PDXP. Pyridoxal 5-prime-phosphate (PLP) is the active form of vitamin B6 that acts as a coenzyme in maintaining biochemical homeostasis. The preferred degradation route from PLP to 4-pyridoxic acid involves the dephosphorylation of PLP by PDXP (Jang et al., 2003 ).
Specifications
| PDXP | |
| Polyclonal | |
| Unconjugated | |
| PDXP | |
| 1600027H05Rik; AB041662; chronophilin; chronophin; CIN; dJ37E16.5; FLJ32703; PDXP; Plp; PLP phosphatase; PLPP; PLP-phosophatase; pyridoxal (pyridoxine, vitamin B6) phosphatase; pyridoxal phosphatase; pyridoxal phosphate phosphatase; Rbp1; reg I binding protein I; reg I-binding protein 1; testicular secretory protein Li 36 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57026 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96GD0 | |
| PDXP | |
| Recombinant protein corresponding to Human PDXP. Recombinant protein control fragment (Product #RP-91385). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction