missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDXK Monoclonal Antibody (1007CT2.1.1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537743
This item is not returnable.
View return policy
Description
PDXK Monoclonal Antibody for Western Blot, IHC (P)
Pyridoxal kinase (PDXK) converts vitamin B6 to pyridoxal-5-phosphate (PLP), an essential cofactor in the intermediate metabolism of amino acids and neurotransmitters. The PDXK gene encodes a 312-amino acid polypeptide, and expression of the cDNA reveals pyridoxal kinase activity. Northern blot analysis revealed that a major 1.5-kb PDXK transcript is expressed in all tissues tested. The expression of PDXK shows circadian oscillations. The expression of Pdxk in mouse liver and brain is regulated by the 3 PAR bZIP transcription factors, Dbp, Hlf, and Tef, which also show circadian oscillations in expression. Mice devoid of all 3 transcription factors show decreased levels of brain PLP, serotonin, and dopamine, and are highly susceptible to frequently lethal generalized spontaneous and audiogenic epilepsies.
Specifications
| PDXK | |
| Monoclonal | |
| Unconjugated | |
| PDXK | |
| 2310036D04Rik; AA119688; C21orf124; C21orf97; C77999; epididymis secretory sperm binding protein Li 1a; HEL-S-1a; Pdxk; Pkh; PNK; PRED79; pyridoxal (pyridoxine, vitamin B6) kinase; pyridoxal kinase; pyridoxamine kinase; pyridoxine kinase; vitamin B6 kinase | |
| Mouse | |
| Protein G | |
| RUO | |
| 8566 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1007CT2.1.1 | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O00764 | |
| PDXK | |
| recombinant protein between 1-312 amino acids from human PDXK. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction