missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDX-1/IPF1 Mouse anti-Human, Clone: PCRP-PDX1-2C11, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07528-100ug
This item is not returnable.
View return policy
Description
PDX-1/IPF1 Monoclonal specifically detects PDX-1/IPF1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.
Specifications
| PDX-1/IPF1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array | |
| Glucose-sensitive factor, IDX-1GSF, Insulin promoter factor 1, insulin promoter factor 1, homeodomain transcription factor, Insulin upstream factor 1, IPF1pancreas/duodenum homeobox protein 1, Islet/duodenum homeobox-1, MODY4IUF1, pancreatic and duodenal homeobox 1, pancreatic-duodenal homeobox factor 1, PDX-1IPF-1, somatostatin transcription factor 1, Somatostatin-transactivating factor 1, STF-1IUF-1 | |
| Recombinant full-length human PDX-1/IPF1 protein | |
| 100 μg | |
| Lipid and Metabolism, Stem Cell Markers | |
| 3651 | |
| Store at 4°C. | |
| IgG2c |
| Flow Cytometry, Immunofluorescence, Peptide Array | |
| PCRP-PDX1-2C11 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu