missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDX-1/IPF1 Antibody (267712) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2419JF525
This item is not returnable.
View return policy
Beskrivning
PDX-1/IPF1 Monoclonal antibody specifically detects PDX-1/IPF1 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Flow Cytometry (Intracellular Staining),Immunocytochemistry
Specifikationer
| PDX-1/IPF1 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| Glucose-sensitive factor, IDX-1GSF, Insulin promoter factor 1, insulin promoter factor 1, homeodomain transcription factor, Insulin upstream factor 1, IPF1pancreas/duodenum homeobox protein 1, Islet/duodenum homeobox-1, MODY4IUF1, pancreatic and duodenal homeobox 1, pancreatic-duodenal homeobox factor 1, PDX-1IPF-1, somatostatin transcription factor 1, Somatostatin-transactivating factor 1, STF-1IUF-1 | |
| E. coli-derived recombinant human PDX-1, Ala91-Arg283, Accession # P52945 | |
| 0.1 mL | |
| Lipid and Metabolism, Stem Cell Markers | |
| 3651 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry | |
| 267712 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering