missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDSS2 Antibody (OTI1D12), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73323CL1
This item is not returnable.
View return policy
Description
PDSS2 Monoclonal antibody specifically detects PDSS2 in Human,Monkey,Mouse,Primate,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
Specifications
| PDSS2 | |
| Monoclonal | |
| CoraFluor 1 | |
| All-trans-decaprenyl-diphosphate synthase subunit 2, bA59I9.3, C6orf210decaprenyl-diphosphate synthase subunit 2, Candidate tumor suppressor protein, chromosome 6 open reading frame 210, Decaprenyl pyrophosphate synthase subunit 2, decaprenyl pyrophosphate synthetase subunit 2, DLP1hDLP1, EC 2.5.1.91, prenyl (decaprenyl) diphosphate synthase, subunit 2, subunit 2 of decaprenyl diphosphate synthase | |
| Full length human recombinant protein of human PDSS2(NP_065114) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Monkey, Mouse, Primate, Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF | |
| OTI1D12 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 57107 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction