missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDSS2 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00057107-B01P
This item is not returnable.
View return policy
Description
PDSS2 Polyclonal antibody specifically detects PDSS2 in Human samples. It is validated for Western Blot
Specifications
| PDSS2 | |
| Polyclonal | |
| Western Blot | |
| NP_065114 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 57107 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| All-trans-decaprenyl-diphosphate synthase subunit 2, bA59I9.3, C6orf210decaprenyl-diphosphate synthase subunit 2, Candidate tumor suppressor protein, chromosome 6 open reading frame 210, Decaprenyl pyrophosphate synthase subunit 2, decaprenyl pyrophosphate synthetase subunit 2, DLP1hDLP1, EC 2.5.1.91, prenyl (decaprenyl) diphosphate synthase, subunit 2, subunit 2 of decaprenyl diphosphate synthase | |
| C6orf210 (NP_065114, 1 a.a. - 399 a.a.) full-length human protein. MNFRQLLLHLPRYLGASGSPRRLWWSPSLDTISSVGSWRGRSSKSPAHWNQVVSEAEKIVGYPTSFMSLRCLLSDELSNIAMQVRKLVGTQHPLLTTARGLVHDSWNSLQLRGLVVLLISKAAGPSSVNTSCQNYDMVSGIYSCQRSLAEITELIHIALLVHRGIVNLNELQSSDGPLKDMQFGNKIAILSGDFLLANACNGLALLQNTKVVELLASALMDLVQGVYHENSTSKESYITDDIGISTWKEQTFLSHGALLAKSCQAAMELAKHDAEVQNMAFQYGKHMAMSHKINSDVQPFIKEKTSDSMTFNLNSAPVVLHQEFLGRDLWIKQIREAQEKGRLDYAKLRERIKAGKGVTSAIDLCRYHGNKALEALESFPPSEARSALENIVFAVTRFS | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction