missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDSS1 Recombinant Rabbit Monoclonal Antibody (K01_1Q56)

Product Code. 30829920
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829920 100 μL
30825179 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829920 Supplier Invitrogen™ Supplier No. MA563376

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

PDSS1 Recombinant Monoclonal Antibody for Western Blot

PDSS1 is an enzyme that elongates the prenyl side-chain of coenzyme Q, or ubiquinone, one of the key elements in the respiratory chain. PDSS1 catalyzes the formation of all trans-polyprenyl pyrophosphates from isopentyl diphosphate in the assembly of polyisoprenoid side chains, the first step in coenzyme Q biosynthesis. The protein may be peripherally associated with the inner mitochondrial membrane, though no transit peptide has been definitively identified to date. Defects in PDSS1 gene are a cause of coenzyme Q10 deficiency.The protein encoded by this gene is an enzyme that elongates the prenyl side-chain of coenzyme Q, or ubiquinone, one of the key elements in the respiratory chain. The gene product catalyzes the formation of all trans-polyprenyl pyrophosphates from isopentyl diphosphate in the assembly of polyisoprenoid side chains, the first step in coenzyme Q biosynthesis. The protein may be peripherally associated with the inner mitochondrial membrane, though no transit peptide has been definitively identified to date. Defects in this gene are a cause of coenzyme Q10 deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDSS1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1Q56
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene PDSS1
Gene Accession No. Q5T2R2
Gene Alias 2610203G20Rik; 2700031G06Rik; All-trans-decaprenyl-diphosphate synthase subunit 1; COenzyme Q (ubiquinone) biosynthesis family member (coq-1)-like; coenzyme Q1 homolog; COQ1; COQ10D2; decaprenyl diphdsphate synthase subunit1; decaprenyl diphosphate synthase subunit 1; decaprenyl pyrophosphate synthase subunit 1; decaprenyl pyrophosphate synthetase subunit 1; decaprenyl-diphosphate synthase subunit 1; DPS; Dps1; hDPS1; hypothetical protein LOC550349; LOW QUALITY PROTEIN: decaprenyl-diphosphate synthase subunit 1; mDLP1; MGC70953; mSPS1; PDSS1; polyprenyl pyrophosphate synthetase; prenyl (decaprenyl) diphosphate synthase, subunit 1; prenyl (solanesyl) diphosphate synthase, subunit 1; prenyl diphosphate synthase, subunit 1; RP13-16H11.3; solanesyl-diphosphate synthase subunit 1; SPS; Sps1; subunit 1 of decaprenyl diphosphate synthase; subunit 1 of solanesyl diphosphate synthase; TPRT; TPT; TPT 1; trans-prenyltransferase; trans-prenyltransferase (TPT); trans-prenyltransferase 1; wu:fc11f12; wu:fd05d05; zgc:112058
Gene Symbols PDSS1
Host Species Rabbit
Immunogen A synthetic peptide of human DPS1.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23590
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.