missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDS5B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18796-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that interacts with the conserved protein complex termed cohesin. The cohesin complex holds together sister chromatids and facilitates accurate chromosome segregation during mitosis and meiosis. This protein is also a negative regulator of cell proliferation and may be a tumor-suppressor gene.Specifications
| PDS5B | |
| Polyclonal | |
| Unconjugated | |
| PDS5B | |
| APRIN, AS3, CG008, KIAA0979, PDS5B, RP1 267P19.1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100710, 23047, 304218 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q4VA53, Q6TRW4, Q9NTI5 | |
| PDS5B | |
| PDS5B Fusion Protein Ag8301 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction