missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDS5B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103601
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PDS5B.
This gene encodes a protein that interacts with the conserved protein complex termed cohesin. The cohesin complex holds together sister chromatids and facilitates accurate chromosome segregation during mitosis and meiosis. This protein is also a negative regulator of cell proliferation and may be a tumor-suppressor gene.
Specifications
| PDS5B | |
| Polyclonal | |
| Unconjugated | |
| PDS5B | |
| AI646570; androgen induced inhibitor of proliferation; androgen-induced proliferation inhibitor; androgen-induced prostate proliferative shutoff associated protein; androgen-induced prostate proliferative shutoff associated protein AS3; androgen-induced prostate proliferative shutoff-associated protein AS3; androgen-induced shutoff 3; Aprin; As3; AW212954; CG008; Kiaa0979; mKIAA0979; PDS5 cohesin associated factor B; PDS5, regulator of cohesion maintenance, homolog B; PDS5, regulator of cohesion maintenance, homolog B (S. cerevisiae); Pds5b; Sister chromatid cohesion protein PDS5 homolog B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 100710, 23047, 304218 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q4VA53, Q6TRW4, Q9NTI5 | |
| PDS5B | |
| A synthesized peptide derived from human PDS5B(Accession Q9NTI5), corresponding to amino acid residues A1356-S1406. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction