missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDS5B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558824
This item is not returnable.
View return policy
Description
Immunogen sequence: ESPESSAIES TQSTPQKGRG RPSKTPSPSQ PKKNVRVGRS KQAATKENDS SEEVDVFQGS SPVDDIPQEE TEEEEVSTVN VRRRSAKRER Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 81%.
This gene encodes a protein that interacts with the conserved protein complex termed cohesin. The cohesin complex holds together sister chromatids and facilitates accurate chromosome segregation during mitosis and meiosis. This protein is also a negative regulator of cell proliferation and may be a tumor-suppressor gene.
Specifications
| PDS5B | |
| Polyclonal | |
| Unconjugated | |
| PDS5B | |
| AI646570; androgen induced inhibitor of proliferation; androgen-induced proliferation inhibitor; androgen-induced prostate proliferative shutoff associated protein; androgen-induced prostate proliferative shutoff associated protein AS3; androgen-induced prostate proliferative shutoff-associated protein AS3; androgen-induced shutoff 3; Aprin; As3; AW212954; CG008; Kiaa0979; mKIAA0979; PDS5 cohesin associated factor B; PDS5, regulator of cohesion maintenance, homolog B; PDS5, regulator of cohesion maintenance, homolog B (S. cerevisiae); Pds5b; Sister chromatid cohesion protein PDS5 homolog B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23047 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NTI5 | |
| PDS5B | |
| Recombinant protein corresponding to Human PDS5B. Recombinant protein control fragment (Product #RP-97918). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction