missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDS5A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542834
This item is not returnable.
View return policy
Description
Peptide sequence: PPAPSKPRRG RRPKSESQGN ATKNDDLNKP INKGRKRAAV GQESPGGLEA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
Nop56p is a yeast nucleolar protein that is part of a complex with the nucleolar proteins Nop58p and fibrillarin. Nop56p is required for assembly of the 60S ribosomal subunit and is involved in pre-rRNA processing. NOL5A is similar in sequence to Nop56p and is also found in the nucleolus. Multiple transcript variants encoding several different isoforms have been found for this gene, but the full-length nature of most of them have not been determined.Nop56p is a yeast nucleolar protein that is part of a complex with the nucleolar proteins Nop58p and fibrillarin. Nop56p is required for assembly of the 60S ribosomal subunit and is involved in pre-rRNA processing. The protein encoded by this gene is similar in sequence to Nop56p and is also found in the nucleolus. Multiple transcript variants encoding several different isoforms have been found for this gene, but the full-length nature of most of them have not been determined.
Specifications
| PDS5A | |
| Polyclonal | |
| Unconjugated | |
| PDS5A | |
| 9030416H16Rik; Cell proliferation-inducing gene 54 protein; E230024D05Rik; KIAA0648; mKIAA0648; PDS5; PDS5 cohesin associated factor A; PDS5, regulator of cohesion maintenance, homolog A; PDS5, regulator of cohesion maintenance, homolog A (S. cerevisiae); PDS5A; PIG54; RGD1307094; SCC112; SCC-112; Sister chromatid cohesion protein 112; sister chromatid cohesion protein PDS5 homolog A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23244 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q29RF7 | |
| PDS5A | |
| Synthetic peptide directed towards the C-terminal of human PDS5A (aa 1262-1311). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction