missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDRG1 Antibody (OTI4F12), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73320PEATT594
This item is not returnable.
View return policy
Description
PDRG1 Monoclonal specifically detects PDRG1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| PDRG1 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| PDRG1 | |
| Full length recombinant protein of human PDRG1 (NP_110442) produced in HEK293T cells. | |
| 0.1 mL | |
| 81572.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI4F12 | |
| Flow Cytometry | |
| C20orf126p53 and DNA damage regulated 1, chromosome 20 open reading frame 126, dJ310O13.3, p53 and DNA-damage regulated 1, PDRGp53 and DNA damage-regulated protein 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction